Learn More
World Precision Instrument Microelectrode Holder (MEH7W)-MEH7W15
SDP
Catalog No. 502521094
Encompass
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.
Catalog No. 50-252-1094 Supplier World Precision Instrument Supplier No. MEH7W15
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.