Learn More
World Precision Instrument Microelectrode Holder (MEH8)-MEH812
SDP
Catalog No. 502521092
Encompass
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.
Catalog No. 50-252-1092 Supplier World Precision Instrument Supplier No. MEH812
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.