Learn More
World Precision Instrument Microelectrode Holder (MEH8)-MEH815
SDP
Catalog No. 502521091
Encompass
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.
Catalog No. 50-252-1091 Supplier World Precision Instrument Supplier No. MEH815
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.