Learn More
World Precision Instrument Microelectrode Holder (MEH8)-MEH820
SDP
Catalog No. 502521090
Encompass
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.
Catalog No. 50-252-1090 Supplier World Precision Instrument Supplier No. MEH820
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.