Learn More
World Precision Instrument Microelectrode Holder (MEH900R)-MEH900R10
SDP
Catalog No. 502538696
Encompass
For use with these WPI Products: SYS-900A
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.
Catalog No. 50-253-8696 Supplier World Precision Instrument Supplier No. MEH900R10
For use with these WPI Products: SYS-900A
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.