Learn More
World Precision Instrument Microelectrode Holder (MEH900S)-MEH900S15
SDP
Catalog No. 502521086
Encompass
For use with these WPI Products: SYS-900A
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.
Catalog No. 50-252-1086 Supplier World Precision Instrument Supplier No. MEH900S15
For use with these WPI Products: SYS-900A
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.