Learn More
World Precision Instrument Microelectrode Holder (MPH1)-MPH115
SDP
Catalog No. 502538680
Encompass
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.
Catalog No. 50-253-8680 Supplier World Precision Instrument Supplier No. MPH115
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.