Learn More
World Precision Instrument Microelectrode Holder (MPH1)-MPH112
SDP
Catalog No. 502538681
Encompass
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.
Catalog No. 50-253-8681 Supplier World Precision Instrument Supplier No. MPH112
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.