Learn More
World Precision Instrument Microelectrode Holder (MPH1)-MPH110
SDP
Catalog No. 502538682
Encompass
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.
Catalog No. 50-253-8682 Supplier World Precision Instrument Supplier No. MPH110
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.