Learn More
World Precision Instrument Microelectrode Holder (MPH3)-MPH315
SDP
Catalog No. 502521074
Encompass
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.
Catalog No. 50-252-1074 Supplier World Precision Instrument Supplier No. MPH315
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.