Learn More
World Precision Instrument Microelectrode Holder (MPH3)-MPH320
SDP
Catalog No. 502538677
Encompass
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.
Catalog No. 50-253-8677 Supplier World Precision Instrument Supplier No. MPH320
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.