Learn More
World Precision Instrument Microelectrode Holder (MPH4)-MPH420
SDP
Catalog No. 502521070
Encompass
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.
Catalog No. 50-252-1070 Supplier World Precision Instrument Supplier No. MPH420
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.