Learn More
World Precision Instrument Microelectrode Holder (MPH4)-MPH410
SDP
Catalog No. 502521073
Encompass
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.
Catalog No. 50-252-1073 Supplier World Precision Instrument Supplier No. MPH410
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.