Learn More
World Precision Instrument Microelectrode Holder (MPH4)-MPH415
SDP
Catalog No. 502521071
Encompass
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.
Catalog No. 50-252-1071 Supplier World Precision Instrument Supplier No. MPH415
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.