Learn More
World Precision Instrument Microelectrode Holder (MPH4)-MPH412
SDP
Catalog No. 502521072
Encompass
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.
Catalog No. 50-252-1072 Supplier World Precision Instrument Supplier No. MPH412
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.