Learn More
World Precision Instrument Microelectrode Holder (MPH6P)-MPH6P15
SDP
Catalog No. 502538675
Encompass
For use with these WPI Products: Piconozzle Kit (5430-XX)
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.
Catalog No. 50-253-8675 Supplier World Precision Instrument Supplier No. MPH6P15
For use with these WPI Products: Piconozzle Kit (5430-XX)
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.