Learn More
World Precision Instrument Microelectrode Holder (MPH6P)-MPH6P10
SDP
Catalog No. 502538676
Encompass
For use with these WPI Products: Piconozzle Kit (5430-XX)
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.
Catalog No. 50-253-8676 Supplier World Precision Instrument Supplier No. MPH6P10
For use with these WPI Products: Piconozzle Kit (5430-XX)
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.