Learn More
World Precision Instrument Microelectrode Holder (MPH6S)-MPH6S15
SDP
Catalog No. 502521065
Encompass
For use with these WPI Products: Piconozzle Kit (5430-XX)
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters.
Catalog No. 50-252-1065 Supplier World Precision Instrument Supplier No. MPH6S15
For use with these WPI Products: Piconozzle Kit (5430-XX)
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters.