Learn More
Cole-Parmer Microelectrode Ph BNC Connector
Catalog No. NC1821214 Shop All Cole-Parmer Products
Encompass_Preferred
Take pH measurements in samples as low as 10 uL, even from filter paper and agarose gel.
Catalog No. NC1821214 Supplier Cole-Parmer Supplier No. 0599300
Take pH measurements in samples as low as 10 uL, even from filter paper and agarose gel.