Learn More
World Precision Instrument Microelectrode Puller
SDP
Catalog No. 502538620
Encompass
PUL-1000 is a microprocessor controlled horizontal puller for making glass micropipettes or microelectrodes used in intracellular recording, patch clamp studies, microperfusion or microinjection. The puller was designed with tight mechanical specifications and precision electronics for complete control of the pulling process and accurate reproducibility. It offers programmable sequences of up to four steps with heating, force, movement and cooling time. This allows graduated cycles for applications like patch clamp recording.
View our Micropipette Puller Comparison Chart
Catalog No. 50-253-8620 Supplier World Precision Instrument Supplier No. PUL1000
Add to Cart
Add to Cart
Request a Quote
PUL-1000 is a microprocessor controlled horizontal puller for making glass micropipettes or microelectrodes used in intracellular recording, patch clamp studies, microperfusion or microinjection. The puller was designed with tight mechanical specifications and precision electronics for complete control of the pulling process and accurate reproducibility. It offers programmable sequences of up to four steps with heating, force, movement and cooling time. This allows graduated cycles for applications like patch clamp recording.
View our Micropipette Puller Comparison Chart