Learn More
Perkin Elmer US LLC MicroFAST SC FAST Probe
Catalog No. 502104686
Encompass_Preferred
This probe has Carbon Fiber support with -28 fittings and a capillary length of 100 cm (after probe). This SC FAST Probe is included in the MicroFAST Line Valve Complete Assembly (N8150400).
Catalog No. 50-210-4686 Supplier Perkin Elmer US LLC Supplier No. N8152679
This probe has Carbon Fiber support with -28 fittings and a capillary length of 100 cm (after probe). This SC FAST Probe is included in the MicroFAST Line Valve Complete Assembly (N8150400).