Learn More
MSC Contec Professional Wet Mop Loop, Clamp Jaw, White Microfiber, 24/Pk
Catalog No. 502818886
Encompass
This wet mop loop features a clamp jaw connection and microfiber head material with a loop style, white mop, and white headband. Made from laundry-free Premira Microfiber, it offers excellent pickup of dirt, hair, and debris. Designed for high-volume spill areas like ORs and labor/delivery, it absorbs more than traditional mops and is brick-packed for optimized storage.
  • Connection type: Clamp jaw
  • Head material: Microfiber
  • Style: Loop
  • Mop color: White
  • Headband color: White
  • Made from laundry-free microfiber
  • Provides excellent pickup of dirt, hair, and small debris
  • Absorbs more than traditional grossing mops
  • Ideal for ORs, high irrigation cases, labor and delivery, and other high volume spill areas
  • Brick-packed for optimized storage
Catalog No. 50-281-8886 Supplier MSC Supplier No. 11570702
This wet mop loop features a clamp jaw connection and microfiber head material with a loop style, white mop, and white headband. Made from laundry-free Premira Microfiber, it offers excellent pickup of dirt, hair, and debris. Designed for high-volume spill areas like ORs and labor/delivery, it absorbs more than traditional mops and is brick-packed for optimized storage.
  • Connection type: Clamp jaw
  • Head material: Microfiber
  • Style: Loop
  • Mop color: White
  • Headband color: White
  • Made from laundry-free microfiber
  • Provides excellent pickup of dirt, hair, and small debris
  • Absorbs more than traditional grossing mops
  • Ideal for ORs, high irrigation cases, labor and delivery, and other high volume spill areas
  • Brick-packed for optimized storage