Learn More
ACL Inc Staticide 4" X 4", 600 Microfiber wipers per bag
Catalog No. NC1841770
Encompass_Preferred
80% Polyester / 20% nylon ultra-low particle and fiber release, ultrasonically sealed edges, high absorbency, durablew strength in wet applications, ISO Class 5 (Class 100).
Catalog No. NC1841770 Supplier ACL Inc Supplier No. 8404MF
80% Polyester / 20% nylon ultra-low particle and fiber release, ultrasonically sealed edges, high absorbency, durablew strength in wet applications, ISO Class 5 (Class 100).