Learn More
Electron Microscopy Sciences Knit Poly/Nylon Microfiber All-Purpose Swabs
SDP
Catalog No. 501930633
Encompass_Preferred
Very clean and durable all-purpose swabs (knit or nonwoven fabric heads) are acid and solvent resistant. Sealed edges build in high strength with no loose fibers or particulates. They are ideal for tough scrubbing around raised shapes. Cleanroom process gives you the lowest ionic, non-volatile residue and particle contaminants. Polypropylene handle.
Catalog No. 50-193-0633 Supplier Electron Microscopy Sciences Supplier No. 7832578
Very clean and durable all-purpose swabs (knit or nonwoven fabric heads) are acid and solvent resistant. Sealed edges build in high strength with no loose fibers or particulates. They are ideal for tough scrubbing around raised shapes. Cleanroom process gives you the lowest ionic, non-volatile residue and particle contaminants. Polypropylene handle.