Learn More
World Precision Instrument MicroFil-MF34G-5
SDP
Catalog No. 50821914
Encompass
Our MicroFil fills micropipettes easily and reliably. Its long and fine tip allows you to start the filling very close to the pipette tip, eliminating both air bubble formation and clogging due to the washing down of dust particles. The transparent amber MicroFil needle is constructed from a combination of plastic and fused silica - no metal components are used. The MicroFil plastic needle may be stored for days with the filling solution inside without clogging. Sold in packages of 5.
Catalog No. 50-821-914 Supplier World Precision Instrument Supplier No. MF34G5
Our MicroFil fills micropipettes easily and reliably. Its long and fine tip allows you to start the filling very close to the pipette tip, eliminating both air bubble formation and clogging due to the washing down of dust particles. The transparent amber MicroFil needle is constructed from a combination of plastic and fused silica - no metal components are used. The MicroFil plastic needle may be stored for days with the filling solution inside without clogging. Sold in packages of 5.