Learn More
MilliporeSigma Microfil™ Funnel Dispenser
Catalog No. MIACFD201 Shop All MilliporeSigma Products
Change view
Click to view available options
:
Funnel Dispenser
Catalog No.
MIACFD201 Funnel Dispenser
1 options
Catalog No. MIACFD201 Supplier MilliporeSigma Supplier No. MIACFD201
Only null left
Add to Cart
Add to Cart
Dispenser for use with 250 mL Microfil™ funnels.
The funnel dispenser for the 250 mL funnel is an additional accessory of the Microfil™ filtration system. The Microfil™ filtration system is a simple and reliable solution for routine microbiological testing. The push-fit design of the Microfil™ funnels seals tightly to manifold supports without the need for clamps or O-rings. The funnels, packed in convenient packs of 25, are loaded to the custom-designed funnel dispenser and can be easily removed as needed. The funnel dispenser is designed to allow rapid dispensing of Microfil™ funnels without compromising sterility.
TRUSTED_SUSTAINABILITY
Specifications
Disposable Reusable
For Use With (Equipment) EZ-Fit Filtration Manifold
Type Funnel Dispenser