Learn More
Sigma Aldrich Fine Chemicals Biosciences Microfil(R) funnel with S-Pak(R) sample volume100 mL, mixed cellulose esters (MCE), pore size 0.22 mum, filter diam. 47 mm, white filter, gridded filter |
Supplier:  Sigma Aldrich Fine Chemicals Biosciences MIGSWG100
Encompass
Microfil(R) funnel with S-Pak(R) sample volume100 mL, mixed cellulose esters (MCE), pore size 0.22 mum, filter diam. 47 mm, white filter, gridded filter |
Catalog No. NC1940300