Learn More
Description
Includes
16-channel manifold for 96- and 384-well microplates, aerosol cover, priming trough self-draining kit
MicroFill is an economical, compact, and reliable dispenser for 24-, 96-, and 384-well microplates. An autoclavable fluid path is ideal for applications that require sterility. The dispense flow rates are easily selected to accommodate sensitive cell-based assays or vigorous dispensing for ELISA
Accurate and precise dispensing are guaranteed, without the time-consuming recalibration, cassette replacement, and maintenance commonly associated with other dispensers. Intuitive, on-board software is easy to use and saves time in instrument setup.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Accuracy | 1μL typical at 5μL, 1μL typical at 20μL, 1% typical at 100μL |
| Product Type | Microplate Dispenser |
| Certifications/Compliance | CE and TUV marked. RoHS compliant. |
| Volume (Metric) | 5 to 6000 μL |
| Increments | 1 μL |
| Interface | 2 x 24 LCD character screen, 25 alphanumeric soft keys |
| Model | AF1000A |
| Operating Conditions | 80% Humidity |
| Plate Type | 24, 96 and 384-well plates |
| Precision | <5% CV typical at 5μL, <2.5% CV typical at 20μL, <1% CV typical at 100μL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
