Learn More
IDEX Health & Science Upchurch MICROFINGER 6-32 PEEK
Catalog No. NC1919495
Encompass_Preferred
Idex MicroTight One-piece Fittings, Packaging Quantity: Each, PEEK, One Piece Fitting, Natural, Overall Length: 0.53 in., 13.7 mm, Overall Width: 0.31 in., 7.87 mm, Pressure: 4000 psi
Catalog No. NC1919495 Supplier IDEX Health & Science Upchurch Supplier No. F125
Idex MicroTight One-piece Fittings, Packaging Quantity: Each, PEEK, One Piece Fitting, Natural, Overall Length: 0.53 in., 13.7 mm, Overall Width: 0.31 in., 7.87 mm, Pressure: 4000 psi