Learn More
Research Products International Corp MICROFLEX 92-134 Gloves, Medium, Case
SDP
Supplier:  Research Products International Corp 036530MCS
Encompass
The MICROFLEX 92-134 disposable nitrile examination glove offers tactility and comfortable protection across numerous applications. The lightweight, soft and stretchy nitrile formulation offers a comfortable fit and feel with the tactile sensitivity needed for fine motor control. Designed with textured fingertips for secure handling of tools and materials.
Catalog No. 50-225-3013