Learn More
World Precision Instrument Microflex Non-Rebreathing Unit
SDP
Catalog No. 502538859
Encompass
The Microflex Non-Rebreathing Unit efficiently delivers anesthetic gas to an animal during a procedure. The unit incorporates our patented valve design that mitigates the gas flow to the nosecone mask. 
Catalog No. 50-253-8859 Supplier World Precision Instrument Supplier No. EZ103A
The Microflex Non-Rebreathing Unit efficiently delivers anesthetic gas to an animal during a procedure. The unit incorporates our patented valve design that mitigates the gas flow to the nosecone mask. 