Learn More
Braintree Scientific Microflex Rebreather w/plastic bed
SDP
Catalog No. 501954771
Encompass
Microflex Breather. This low profile breather is suitable for work under microscopes and other applications where a smaller breather is required. Also available in composite plastic for MRI use. Breather is supplied with two nosecone sizes - customized for mice, rats and other animals weighing up to 500 grams. Tight fitting nosecones eliminate gas leakage and protect personnel from gas exposure. Stainless steelversion includes heating elements for maintaining animal temperature during procedures. **Sold for research purposes only and not for human use**
Catalog No. 50-195-4771 Supplier Braintree Scientific Supplier No. EZ103A
Microflex Breather. This low profile breather is suitable for work under microscopes and other applications where a smaller breather is required. Also available in composite plastic for MRI use. Breather is supplied with two nosecone sizes - customized for mice, rats and other animals weighing up to 500 grams. Tight fitting nosecones eliminate gas leakage and protect personnel from gas exposure. Stainless steelversion includes heating elements for maintaining animal temperature during procedures. **Sold for research purposes only and not for human use**