Learn More
Carson MicroFlip with Prepared Slides
SDPSCI_ED
Catalog No. S01592 Shop All Carson Optical Products
Change view
Click to view available options
Magnification:
100 to 250x
Catalog No. Magnification
S01592 100 to 250x
1 options
This item is not returnable. View return policy
Catalog No. S01592 Supplier Carson Supplier No. MP250BUN
Add to Cart
Add to Cart
This item is not returnable. View return policy
The MicroFlip from Carson is a pocket microscope with LED and UV lights and a flip-down slide base smartphone clip.
  • Includes 24 prepared slides and a smartphone digiscoping clip that allows you to take pictures and videos using your phone's camera app.
  • The magnification ranges from 100x to 250x power, and there's a built-in UV flashlight for detecting counterfeit currency and other things.
  • The digiscoping clip works with most smartphone models, including iPhone and Samsung, and it's powered by 1 AA battery (not included).
  • The bundled version also includes a starter slide with cover slip and a wrist strap for portability.
Specifications
Magnification 100 to 250x
Product Type Microscope
Material Acrylic