Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Paddle Wheel Micro-Flo Rate and Total Flowmeter, 1/4"" NPT(F); 5000 mL/min
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX3255009
Encompass
These economical flowmeters are capable of very low flow rates. The display is programmable for preselected factory settings or can be field calibrated for high accuracy in a specific application. Displays units in milliliters, ounces, liters, or gallons; displays time in minutes, hours, or days. Display can be programmed for up to four decimal places. All units are supplied with a 115 VAC/DC power supply.
Catalog No. NC1531548