Learn More
World Precision Instrument MicroFlow - Flow-through housed oxygen microsensor
SDP
Catalog No. 502521407
Encompass
The MicroFlow fiber optic oxygen sensor is a miniaturized fiber optic chemical sensor optimized for fast response time (t90 < 1 sec in gases, < 5 sec in liquids). The tiny probe has a tip size of 50 µm and is integrated in a T-shape flow cell for easy connection via Luer-Lock adapters to external tubings. Liquids (like water, blood, etc.) can be pumped through the cell.
Catalog No. 50-252-1407 Supplier World Precision Instrument Supplier No. 501657
The MicroFlow fiber optic oxygen sensor is a miniaturized fiber optic chemical sensor optimized for fast response time (t90 < 1 sec in gases, < 5 sec in liquids). The tiny probe has a tip size of 50 µm and is integrated in a T-shape flow cell for easy connection via Luer-Lock adapters to external tubings. Liquids (like water, blood, etc.) can be pumped through the cell.