Learn More
Elemental Scientific Inc MICROFLOW PFA-ST SELF-ASPIRATI
SDP
Supplier:  Elemental Scientific Inc ES2040NFNC
Encompass_Preferred
MicroFlow PFA-ST self-aspirating nebulizer with external ¼-28 threadedconnector (No capillaries or gas fittings included)
Catalog No. NC2156405