Learn More
World Precision Instrument Microforge with Digital Controller-DMF1000-2
SDP
Catalog No. 502538867
Encompass
The DMF1000 is a microprocessor-controlled microforge offering unmatched performance. Designed for fabrication of both small patch clamp glass pipettes and larger injection pipettes, the DMF1000 should find many uses in the laboratory. The DMF1000 is based on a design similar to that first used in WPI’s extremely popular microforge model, the MF200. The extensive improvements incorporated into the DMF1000 greatly increase its versatility and performance, making it one of the most powerful microforges on the market.
Catalog No. 50-253-8867 Supplier World Precision Instrument Supplier No. DMF10002
Add to Cart
Add to Cart
Request a Quote
The DMF1000 is a microprocessor-controlled microforge offering unmatched performance. Designed for fabrication of both small patch clamp glass pipettes and larger injection pipettes, the DMF1000 should find many uses in the laboratory. The DMF1000 is based on a design similar to that first used in WPI’s extremely popular microforge model, the MF200. The extensive improvements incorporated into the DMF1000 greatly increase its versatility and performance, making it one of the most powerful microforges on the market.