Learn More
Beckman Coulter Beckman Coulter Microfuge 20 w FA241.5P
Catalog No. 501631968
Encompass_Preferred
Microfuge 20 microcentrifuge, IVD, w/FA241.5P fixed angle rotor, 120 V/60 Hz
This item is not returnable. View return policy
Catalog No. 50-163-1968 Supplier Beckman Coulter Supplier No. B30137
Microfuge 20 microcentrifuge, IVD, w/FA241.5P fixed angle rotor, 120 V/60 Hz