Learn More
Vee Gee Heathrow Scientific Microfuge Tube 0.2mL NatPK1000
Catalog No. 502886387
Encompass_Preferred
These microfuge tubes feature attached flat caps for easy one-handed use.* Attached flat caps for easy one-handed use* 0.2 mL is suitable for thermal cyclers* 100% polypropylene* Certified DNase, RNase & Pyrogen Free
Catalog No. 50-288-6387 Supplier Vee Gee Supplier No. HS4423
These microfuge tubes feature attached flat caps for easy one-handed use.* Attached flat caps for easy one-handed use* 0.2 mL is suitable for thermal cyclers* 100% polypropylene* Certified DNase, RNase & Pyrogen Free