Learn More
Vee Gee Microfuge Tube 1.5mL Nat PK500
Catalog No. 502886378
Encompass_Preferred
These microfuge tubes feature attached flat caps for easy one-handed use.* Attached flat caps for easy one-handed use* 0.5 and 1.5 mL versions have a frosted writing area and a syringe pierceable cap* 100% polypropylene* Certified DNase, RNase & Pyrogen Free
Catalog No. 50-288-6378 Supplier Vee Gee Supplier No. HS4323
These microfuge tubes feature attached flat caps for easy one-handed use.* Attached flat caps for easy one-handed use* 0.5 and 1.5 mL versions have a frosted writing area and a syringe pierceable cap* 100% polypropylene* Certified DNase, RNase & Pyrogen Free