Learn More
Vee Gee Heathrow Scientific Microfuge Tube 1.5mL NatPK1000
Catalog No. 502886386
Encompass_Preferred
These microfuge tubes feature attached flat caps for easy one-handed use.* Attached flat caps for easy one-handed use* 0.5 and 1.5 mL versions have a frosted writing area and a syringe pierceable cap* 100% polypropylene
Catalog No. 50-288-6386 Supplier Vee Gee Supplier No. HS4325
These microfuge tubes feature attached flat caps for easy one-handed use.* Attached flat caps for easy one-handed use* 0.5 and 1.5 mL versions have a frosted writing area and a syringe pierceable cap* 100% polypropylene