Learn More
Andwin Scientific MICROFUGE TUBE CLEAR/250/PK
SDP
Catalog No. NC9262440
Encompass_Preferred
The Slick Disposable Microcentrifuge Tubes are ideal for use with rDNA and viscous proteins. The tubes are nonsterile, autoclavable, and nonsticking. This product also has feature flat caps that provide leakproof secure snap. There are also frosted a
Catalog No. NC9262440 Supplier Andwin Scientific Supplier No. C33041
The Slick Disposable Microcentrifuge Tubes are ideal for use with rDNA and viscous proteins. The tubes are nonsterile, autoclavable, and nonsticking. This product also has feature flat caps that provide leakproof secure snap. There are also frosted a