Learn More
Cytiva (Formerly Pall Lab) Laboratory Manifold MicroFunnel™ Adapter (3/pkg)
Supplier:  Cytiva 4890
Encompass
Accessory adapter for MicroFunnel filter funnels for microbiology manifold Lab designed for flexibility and a reduction in cross-contamination with interchangeable adapters for most funnels and coupling device to double testing capacity while still enabling sterilization in standard autoclaves
Catalog No. NC1862853