Learn More
Sciencell Research Laboratories MICROGLIA MEDIUM
Catalog No. NC0180234
Encompass
Microglia medium is a complete medium designed for optimal growth of normal human brain-derived microglia in vitro. It is shipped as a kit with microglia growth supplement and Pen/Strep solution.
Catalog No. NC0180234 Supplier Sciencell Research Laboratories Supplier No. 1901
Microglia medium is a complete medium designed for optimal growth of normal human brain-derived microglia in vitro. It is shipped as a kit with microglia growth supplement and Pen/Strep solution.