Learn More
International Enviroguard™ MicroGuard MP Microporous Apron with Ties in Back
Catalog No. 17184079 Shop All International Enviroguard Systems Products
Change view
Click to view available options
Closure Type:
Waist Ties
Catalog No. Closure Type
17-184-079 Waist Ties
1 options
Catalog No. 17-184-079 Supplier International Enviroguard™ Supplier No. 802448
Add to Cart
Add to Cart
MicroGuard MP disposable aprons are produced with state-of-the-art lamination technology and have a microporous film.
  • Soft, non-woven fabric offers breathability while also allowing moisture to escape so you stay cooler throughout long shifts
  • Apron provides frontal protection along with an ideal liquid and particulate barrier
  • Tie strings fasten in the back to keep them out of the way while working, also allowing you to adjust the apron to better fit your size
  • Environmentally friendly; can be recycled after use
Specifications
Type Protective Apron, Bib Apron
Size Universal
Material Microporous Laminate, Polypropylene
Color Yellow
Disposable Single-use
Closure Type Waist Ties
Sterility Non-sterile
Latex Free Yes
No. of Pockets None
Resistance Chemical
Dimensions (L x W) 46 x 28 in.
Length (English) 46 in.
Width (English) 28 in.
No. per Case 100 Aprons/Cs.
Packaging Type Case
Style Bib-style design
Seam Type Seamless