Learn More
International Enviroguard™ MicroGuard MP Microporous Shoe Cover with Elastic Closure
Catalog No. 17184070 Shop All International Enviroguard Systems Products
Change view
Click to view available options
Closure Type:
Elastic Top
Catalog No. Closure Type
17-184-070 Elastic Top
1 options
Catalog No. 17-184-070 Supplier International Enviroguard™ Supplier No. 8101
Add to Cart
Add to Cart
Using state-of-the-art lamination technology, MicroGuard MP microporous shoe covers combine a soft, non-woven fabric with a microporous film.
  • Lightweight, tear-resistant liquid and particulate barrier helps safeguard from blood penetration, various industrial chemicals, along with dust, dirt, and grime
  • Breathable fabric allows moisture to escape, so your feet are more cool and comfortable even as you sweat
  • Shoe covers come with a strong elastic closure at the top to keep them in place
  • Environmentally friendly; can be recycled after use
  • Passes ASTM F1670 testing for blood penetration
Specifications
Size Large
Material Multi-layer Barrier Laminate Fabric, Spunbond Polypropylene
Latex Free Yes
Color White
Disposable Single-use
Sterility Non-sterile
Closure Type Elastic Top
Sole Type Standard
Seam Type Serged
Resistance Abrasion, Static, Liquid, Grime, Dry Particulates
Product Type Shoe Covers