Learn More
International Enviroguard™ MicroGuard MP Microporous Shoe Cover with Non-skid Sole and Elastic Closure
Catalog No. 17184068 Shop All International Enviroguard Systems Products
Change view
Click to view available options
Size:
Large
X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-184-068 Large
17-184-066 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-184-068 Supplier International Enviroguard™ Supplier No. 8103
Add to Cart
Add to Cart
Using state-of-the-art lamination technology, MicroGuard MP microporous shoe covers combine a soft, non-woven fabric with a microporous film to protect against spills and splashes.
  • Shoe covers are designed with a non-skid sole that helps reduce the risk of slips or falls
  • A tear-resistant liquid and particulate barrier helps safeguard from blood penetration, various industrial chemicals, along with dust, dirt, and grime
  • Breathable fabric allows moisture to escape, so your feet stay cool and comfortable even as you sweat
  • Strong elastic closure on top helps keep these durable shoe covers in place, so your shoes and feet are protected throughout your workday
  • Environmentally friendly; can be recycled after use
  • Made of lightweight, tear-resistant polypropylene
  • Passes ASTM F1670 testing for blood penetration
Specifications
Size Large
Material Multi-layer Barrier Laminate Fabric, Spunbond Polypropylene
Latex Free Yes
Color White
Disposable Single-use
Sterility Non-sterile
Closure Type Elastic Top
Sole Type Anti-skid
Seam Type Serged
Resistance Static, Grime, Dry Particulates, Abrasion, Liquid
Product Type Shoe Covers