Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

International Enviroguard™ MicroGuard MP Microporous Sleeves with Elastic Closures

Catalog No. 17184071 Shop All International Enviroguard Systems Products
Change view
Click to view available options
Color:
White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17-184-071 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-184-071 Supplier International Enviroguard™ Supplier No. 8065
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Using state-of-the-art lamination technology, MicroGuard MP microporous 18 in. sleeves combine a soft, non-woven fabric with a microporous film.

  • Lightweight, tear-resistant liquid and particulate barrier helps safeguard from blood penetration, various industrial chemicals, along with dust, dirt, and grime
  • Breathable fabric allows moisture to escape, so you're more cool and comfortable even as you sweat
  • Generous 18 in. length helps cover more of your arms to decrease the risk of exposing skin or underlying clothing to workplace contaminants
  • Secure elastic closures on both ends help keep sleeves in place even with repetitive movements, allowing you to stay on task without having to continually readjust your sleeves throughout your work day
  • Environmentally friendly; can be recycled after use
  • Passes ASTM F1670 testing for blood penetration

Specifications

Size Universal
Product Type Chemical-resistant Sleeve
Material Multi-layer Barrier Laminate Fabric
Color White
Sterility Non-sterile
Style Elastic at Both Ends
Disposable Single-use
Seam Type Serged
Latex Free Yes
Length (English) 18 in.
Length (Metric) 45.7 cm
Width (English) 7.88 in.
Width (Metric) 20 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.