Learn More
Electron Microscopy Sciences Microheat Hot Plate Model MH4, 12V DC
SDP
Catalog No. 50996884
Encompass
The microheatheat stabilized small hot plate combined with power supply is ideal for thermoplastic gluing of cross-sectional samples for electron microscopy under stereo optical microscope. The Microheat makes the embedding procedure easier and helps ensure bubble-free sticking. Due to its small dimensions it can be easily placed on the specimen table of the stereo optical microscope.
Catalog No. 50-996-884 Supplier Electron Microscopy Sciences Supplier No. 55110MH2
The microheatheat stabilized small hot plate combined with power supply is ideal for thermoplastic gluing of cross-sectional samples for electron microscopy under stereo optical microscope. The Microheat makes the embedding procedure easier and helps ensure bubble-free sticking. Due to its small dimensions it can be easily placed on the specimen table of the stereo optical microscope.