Learn More
Medchemexpress LLC HY-N7595 1mg Medchemexpress, Microhelenin C CAS:63569-07-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN75951MG
Encompass_Preferred
Medchemexpress, HY-N7595 1mg Microhelenin C CAS:63569-07-3 Microhelenin C is a sesquiterpene lactone that can be isolated from Helenium microcephalum, with antitumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3001